missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GLP-1R / GLP1R Protein
A DNA sequence encoding the Mus musculus (Mouse) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_067307.2 |
---|---|
Gene ID (Entrez) | 14652 |
Name | GLP-1R / GLP1R Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16030750
|
enQuireBio™
QP6091-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050750
|
enQuireBio™
QP6091-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16040750
|
enQuireBio™
QP6091-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060750
|
enQuireBio™
QP6091-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070750
|
enQuireBio™
QP6091-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080750
|
enQuireBio™
QP6091-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_067307.2 | |
GLP-1R / GLP1R Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
14652 | |
Research Use Only | |
Glp1r | |
Recombinant Protein | |
E. coli | |
GPRPQGTTVS LSETVQKWRE YRRQCQRFLT EAPLLATGLF CNRTFDDYAC WPDGPPGSFV NVSCPWYLPW ASSVLQGHVY RFCTAEGLWL HKDNSSLPWR DLSECEESKR GERNFPEEQL LSLY | |
Greater than 80.4% as determined by SDS-PAGE. |