missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GLP-1R / GLP1R Protein

A DNA sequence encoding the Mus musculus (Mouse) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6091-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030750

  • £560.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_067307.2
GLP-1R / GLP1R Protein
Research Use Only
Glp1r
Recombinant Protein
E. coli
GPRPQGTTVS LSETVQKWRE YRRQCQRFLT EAPLLATGLF CNRTFDDYAC WPDGPPGSFV NVSCPWYLPW ASSVLQGHVY RFCTAEGLWL HKDNSSLPWR DLSECEESKR GERNFPEEQL LSLY
Greater than 80.4% as determined by SDS-PAGE.
14652
100 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers