missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GLP-1R / GLP1R Protein

A DNA sequence encoding the Mus musculus (Mouse) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6091-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060750

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_067307.2
GLP-1R / GLP1R Protein
Research Use Only
Glp1r
Recombinant Protein
E. coli
GPRPQGTTVS LSETVQKWRE YRRQCQRFLT EAPLLATGLF CNRTFDDYAC WPDGPPGSFV NVSCPWYLPW ASSVLQGHVY RFCTAEGLWL HKDNSSLPWR DLSECEESKR GERNFPEEQL LSLY
Greater than 80.4% as determined by SDS-PAGE.
14652
200 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers