missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GLP-1R / GLP1R Protein
A DNA sequence encoding the Mus musculus (Mouse) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6091-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_067307.2 | |
GLP-1R / GLP1R Protein | |
Research Use Only | |
Glp1r | |
Recombinant Protein | |
E. coli | |
GPRPQGTTVS LSETVQKWRE YRRQCQRFLT EAPLLATGLF CNRTFDDYAC WPDGPPGSFV NVSCPWYLPW ASSVLQGHVY RFCTAEGLWL HKDNSSLPWR DLSECEESKR GERNFPEEQL LSLY | |
Greater than 80.4% as determined by SDS-PAGE. |
14652 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |