Learn More
Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.
£89.35 - £417.18
Specifications
Antigen | Epithelial Specific Antigen/Ep-CAM Ab-7 |
---|---|
Clone | MOC-31 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12670217
|
Epredia™
MS-1825-S1 |
Concentrated | 500 μL |
£305.00
500µL |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12660217
|
Epredia™
MS-1825-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12650217
|
Epredia™
MS-1825-S |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12630217
|
Epredia™
MS-1825-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Host Species: Mouse
Clone: MOC-31
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)
Molecular Weight: 40 kDa
Positive Control: Colon CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Epithelial Specific Antigen/Ep-CAM Ab-7 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
IgG2a κ |
MOC-31 | |
Monoclonal | |
Mouse | |
Cancer and Tumor Biology | |
Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1) | |
Primary |