missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1825-S1

Code : LD

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12670217

  • £305.00 / 500µL
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Host Species: Mouse

Clone: MOC-31

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)

Molecular Weight: 40 kDa

Positive Control: Colon CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epithelial Specific Antigen/Ep-CAM Ab-7
Monoclonal
Unconjugated
Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)
500 μL
Cancer and Tumor Biology
Human
IgG2a κ
Immunohistochemistry (Paraffin)
MOC-31
Mouse
Colon CA
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers