Learn More
Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1825-S
Code : LD
Additional Details : Weight : 0.00250kg
Description
Host Species: Mouse
Clone: MOC-31
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)
Molecular Weight: 40 kDa
Positive Control: Colon CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Epithelial Specific Antigen/Ep-CAM Ab-7 | |
Monoclonal | |
Unconjugated | |
Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1) | |
1 mL | |
Cancer and Tumor Biology | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
MOC-31 | |
Mouse | |
Colon CA | |
RUO | |
Primary | |
Concentrated |