Learn More
Epredia™ Lab Vision™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-7, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1825-R7
Description
Host Species: Mouse
Clone: MOC-31
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1)
Molecular Weight: 40 kDa
Positive Control: Colon CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| Epithelial Specific Antigen/Ep-CAM Ab-7 | |
| Monoclonal | |
| Unconjugated | |
| Neuraminidase treated cells from a variant small cell lung carcinoma cell line (GLS-1) | |
| RUO | |
| Primary | |
| Prediluted |
| Immunohistochemistry (Paraffin) | |
| MOC-31 | |
| Mouse | |
| 7 mL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.