missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SOURCE™ 15Q
Strong anion exchanger with high chemical stability for wide range of working conditions with good resistance to cleaning conditions at high pH
£318.00 - £663.00
Specifications
Buffer | 20mM N-methylpiperazine, 20mM Piperazine, 20mM Bis-Tris, 20mM Triethanolamine, 20mM Tris, 20mM Diethanolamine, 20mM Ethanolamine, Glycine, 20mM 1,3-diamino-propane, 20mM Piperidine |
---|---|
Product Type | Source 15Q chromato graphy media |
Height (English) | 3.93 in. |
Height (Metric) | 100mm |
Flow Rate | 1800cm/hr. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11540804
|
Cytiva
17-0947-20 |
10mL |
£318.00
10mL |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
11530804
|
Cytiva
17-0947-01 |
50mL |
£663.00
50mL |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
- Bioprocess medium
- Ligand: quaternary ammonium
- Average particle size: 15μm
- Matrix: polystyrene/divinylbenzene
- pH stability working range: 2 to 12
- For polishing steps in both research and industrial applications
- Available in 10mL and 50mL bottles
Specifications
20mM N-methylpiperazine, 20mM Piperazine, 20mM Bis-Tris, 20mM Triethanolamine, 20mM Tris, 20mM Diethanolamine, 20mM Ethanolamine, Glycine, 20mM 1,3-diamino-propane, 20mM Piperidine | |
3.93 in. | |
1800cm/hr. | |
5 bar | |
15μm | |
4° to 30°C | |
15Q Chromatography Media |
Source 15Q chromato graphy media | |
100mm | |
Quaternary Ammonium | |
2 to 12 | |
Polystyrene/Divinylbenzene | |
Polishing steps in both research and industrial applications |