missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SOURCE™ 15Q

Strong anion exchanger with high chemical stability for wide range of working conditions with good resistance to cleaning conditions at high pH

Brand:  Cytiva 17-0947-01

Code : GE

Additional Details : Weight : 0.13800kg


 View more versions of this product

Product Code. 11530804

  • £663.00 / 50mL
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  • Bioprocess medium
  • Ligand: quaternary ammonium
  • Average particle size: 15μm
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 12
  • For polishing steps in both research and industrial applications
  • Available in 10mL and 50mL bottles

Specifications

Specifications

20mM N-methylpiperazine, 20mM Piperazine, 20mM Bis-Tris, 20mM Triethanolamine, 20mM Tris, 20mM Diethanolamine, 20mM Ethanolamine, Glycine, 20mM 1,3-diamino-propane, 20mM Piperidine
50mL
100mm
Quaternary Ammonium
2 to 12
Polystyrene/Divinylbenzene
Polishing steps in both research and industrial applications
Source 15Q chromato graphy media
3.93 in.
1800cm/hr.
5 bar
15μm
4° to 30°C
15Q Chromatography Media
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SOURCE 15Q, 50 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers