missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SOURCE™ 15Q

Product Code. 11540804 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11540804 10 mL 10mL
11530804 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11540804 Supplier Cytiva Supplier No. 17094720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strong anion exchanger with high chemical stability for wide range of working conditions with good resistance to cleaning conditions at high pH

  • Bioprocess medium
  • Ligand: quaternary ammonium
  • Average particle size: 15μm
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 12
  • For polishing steps in both research and industrial applications
  • Available in 10mL and 50mL bottles

TRUSTED_SUSTAINABILITY

Specifications

Buffer 20mM N-methylpiperazine, 20mM Piperazine, 20mM Bis-Tris, 20mM Triethanolamine, 20mM Tris, 20mM Diethanolamine, 20mM Ethanolamine, Glycine, 20mM 1,3-diamino-propane, 20mM Piperidine
Product Type Source 15Q chromato graphy media
Quantity 10 mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate 1800 cm/hr.
Ligand Type Quaternary Ammonium
Max. Pressure 5 bar
pH Range 2 to 12
Particle Size 15μm
Support Type Polystyrene/Divinylbenzene
Content And Storage 4° to 30°C
For Use With (Application) Polishing steps in both research and industrial applications
Matrix 15Q Chromatography Media
Show More Show Less
compliance-icons
Produktidentifikator
  • SOURCE 15Q, 10 ml
Signalwort
  • Warning
Gefahrenkategorie
  • Flammable liquid Category 3
Gefahrenhinweis
  • H226-Flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.