Learn More
DWK Life Sciences Rubber Adapter, from EPDM, for Filter Crucible
The rubber sleeve is required to achieve gas tightness during vacuum filtration.
£20.20 - £26.45
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
11810720
|
DWK Life Sciences
292013109 |
49 mm |
£26.45
Pack of 10 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11800720
|
DWK Life Sciences
292012601 |
41 mm |
£25.05
Pack of 10 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
11890710
|
DWK Life Sciences
292012104 |
33 mm |
£24.20
Pack of 10 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
11880710
|
DWK Life Sciences
292011408 |
26 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
In order to be able to connect a filter and a filtering flask together, a rubber sleeve is required which protects both glass parts from damage on the one hand and connects them gas-tight on the other hand. This gas tightness is required to perform a vacuum filtration.
- For tight closure in vacuum filtration applications
- For filter crucibles
Specifications
10/Pk. | |
Adapter |