Learn More
DWK Life Sciences Rubber Adapter, from EPDM, for Filter Crucible
The rubber sleeve is required to achieve gas tightness during vacuum filtration.
Brand: DWK Life Sciences 292012104
Code : NEW
Additional Details : Weight : 0.06000kg
Description
In order to be able to connect a filter and a filtering flask together, a rubber sleeve is required which protects both glass parts from damage on the one hand and connects them gas-tight on the other hand. This gas tightness is required to perform a vacuum filtration.
- For tight closure in vacuum filtration applications
- For filter crucibles
Specifications
10/Pk. | |
suitable funnel adapter Cat. no. 24 316 22 06 | |
Adapter | |
33 mm |