Learn More
DWK Life Sciences Rubber Adapter, from EPDM, for Filter Crucible
The rubber sleeve is required to achieve gas tightness during vacuum filtration.
Brand:  DWK Life Sciences 292012601
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11800720
  • £25.05 / Pack of 10
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
In order to be able to connect a filter and a filtering flask together, a rubber sleeve is required which protects both glass parts from damage on the one hand and connects them gas-tight on the other hand. This gas tightness is required to perform a vacuum filtration.
  • For tight closure in vacuum filtration applications
  • For filter crucibles
Specifications
Specifications
10/Pk.
suitable funnel adapter Cat. no. 24 316 26 09
Adapter
41 mm