missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Glass Liner
For high or low molecular weight compounds
POA - £349.69
Specifications
Length (Metric) | 7.85 cm |
---|---|
Diameter (Metric) Inner | 0.4 cm |
Diameter (Metric) Outer | 0.63 cm |
For Use With (Equipment) | Agilent GCs Equipped with Split/Splitless Inlets |
Liner Material | Borosilicate Glass |
Product Code | Brand | Product Type Specs | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type Specs | Price | Quantity & Availability | |||||
12912639
|
Restek™
20710 |
IP Cup Splitter Inlet Liner |
£239.58
Pack of 5 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
10145827
|
Restek™
20710 |
IP Cup Splitter Inlet Liner |
£239.58
Pack of 5 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
12942639
|
Restek™
20710-209.5 |
IP Cup Splitter Inlet Liner |
£302.50
Pack of 5 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
12952639
|
Restek™
20710-210.5 |
Cup Splitter Inlet Liner |
£332.75
Pack of 5 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
12962639
|
Restek™
20710-211.5 |
Cup Splitter Inlet Liner |
£349.69
Pack of 5 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
12922629
|
Restek™
20709 |
IP Cup Splitter Inlet Liner |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12932629
|
Restek™
20709-200.1 |
IP Cup Splitter Inlet Liner |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12922639
|
Restek™
20710-200.5 |
IP Cup Splitter Inlet Liner |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Increased sample residence time in the liner for homogeneous vaporization
- 4mm I.D.
- Available with or without packing
Specifications
7.85 cm | |
0.63 cm | |
Borosilicate Glass |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |