missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Glass Liner
For high or low molecular weight compounds
Brand: Restek™ 20710-200.5
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Increased sample residence time in the liner for homogeneous vaporization
- 4mm I.D.
- Available with or without packing
Specifications
Deactivated Wool | |
5/Pk. | |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |
IP Cup Splitter Inlet Liner | |
7.85 cm | |
0.63 cm | |
Borosilicate Glass |