missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Glass Liner

For high or low molecular weight compounds

Brand:  Restek™ 20710-209.5

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12942639

  • £302.50 / Pack of 5
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

  • Increased sample residence time in the liner for homogeneous vaporization
  • 4mm I.D.
  • Available with or without packing

Specifications

Specifications

CarboFrit
5/Pk.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets
IP Cup Splitter Inlet Liner
7.85 cm
0.63 cm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions