missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Coagulation factor XII Protein
A DNA sequence encoding the Sus scrofa (Pig) Coagulation factor XII, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_999407.1 |
---|---|
Gene ID (Entrez) | 397474 |
Name | Coagulation factor XII Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939319
|
enQuireBio™
QP6008-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959319
|
enQuireBio™
QP6008-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949319
|
enQuireBio™
QP6008-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969319
|
enQuireBio™
QP6008-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979319
|
enQuireBio™
QP6008-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15989319
|
enQuireBio™
QP6008-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_999407.1 | |
Coagulation factor XII Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
397474 | |
Research Use Only | |
F12 | |
Recombinant Protein | |
E. coli | |
IPPWKDPRKH KVMASEHTVV LTVTGEPCHF PFQYYRQLYY KCIQRGQRGP RPWCATTPNF EKDQRWAYCL EPMKVKDHCN KGNPCQKGGT CVNMPNGPHC ICPDHFTGKH CQKEKCFEPQ FLQFFQENEI WHRFEPAGVS KCQCKGPKAQ CKPVASQVCS TNPCLNGGSC LQTEGHRLCR CPTGYAGRLC DVDLKERCYS DRGLSYRGMA QTTLSGAPCQ PWASEATYWN MTAEQALNWG LGDHAFCRNP DNDTRPWCFV WRGDQLSWQY CRLARCQAPI GEAPPILTPT QSPSEHQDSP LLSREPQPTT QTPSQNLTSA WCAPPEQRGP LPSAGLVGCG QRLRKRLSSL NR | |
Greater than 90% as determined by SDS-PAGE. |