missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig Coagulation factor XII Protein
A DNA sequence encoding the Sus scrofa (Pig) Coagulation factor XII, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6008-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_999407.1 | |
Coagulation factor XII Protein | |
Research Use Only | |
F12 | |
Recombinant Protein | |
E. coli | |
IPPWKDPRKH KVMASEHTVV LTVTGEPCHF PFQYYRQLYY KCIQRGQRGP RPWCATTPNF EKDQRWAYCL EPMKVKDHCN KGNPCQKGGT CVNMPNGPHC ICPDHFTGKH CQKEKCFEPQ FLQFFQENEI WHRFEPAGVS KCQCKGPKAQ CKPVASQVCS TNPCLNGGSC LQTEGHRLCR CPTGYAGRLC DVDLKERCYS DRGLSYRGMA QTTLSGAPCQ PWASEATYWN MTAEQALNWG LGDHAFCRNP DNDTRPWCFV WRGDQLSWQY CRLARCQAPI GEAPPILTPT QSPSEHQDSP LLSREPQPTT QTPSQNLTSA WCAPPEQRGP LPSAGLVGCG QRLRKRLSSL NR | |
Greater than 90% as determined by SDS-PAGE. |
397474 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |