missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Coagulation factor XII Protein

A DNA sequence encoding the Sus scrofa (Pig) Coagulation factor XII, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6008-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959319

  • £2480.00 / 1mg
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_999407.1
Coagulation factor XII Protein
Research Use Only
F12
Recombinant Protein
E. coli
IPPWKDPRKH KVMASEHTVV LTVTGEPCHF PFQYYRQLYY KCIQRGQRGP RPWCATTPNF EKDQRWAYCL EPMKVKDHCN KGNPCQKGGT CVNMPNGPHC ICPDHFTGKH CQKEKCFEPQ FLQFFQENEI WHRFEPAGVS KCQCKGPKAQ CKPVASQVCS TNPCLNGGSC LQTEGHRLCR CPTGYAGRLC DVDLKERCYS DRGLSYRGMA QTTLSGAPCQ PWASEATYWN MTAEQALNWG LGDHAFCRNP DNDTRPWCFV WRGDQLSWQY CRLARCQAPI GEAPPILTPT QSPSEHQDSP LLSREPQPTT QTPSQNLTSA WCAPPEQRGP LPSAGLVGCG QRLRKRLSSL NR
Greater than 90% as determined by SDS-PAGE.
397474
1 mg
Not determined.
Partial (see sequence information for more details).
Pig
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers