missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nlrp3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nlrp3, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_665826.1 |
---|---|
Gene ID (Entrez) | 216799 |
Name | Nlrp3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16081201
|
enQuireBio™
QP7856-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001211
|
enQuireBio™
QP7856-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091201
|
enQuireBio™
QP7856-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011211
|
enQuireBio™
QP7856-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021211
|
enQuireBio™
QP7856-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031211
|
enQuireBio™
QP7856-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_665826.1 | |
Nlrp3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
216799 | |
Research Use Only | |
Nlrp3 | |
Recombinant Protein | |
E. coli | |
MTSVRCKLAQ YLEDLEDVDL KKFKMHLEDY PPEKGCIPVP RGQMEKADHL DLATLMIDFN GEEKAWAMAV WIFAAINRRD LWEKAKKDQP EWNDTCTSHS SMVCQEDSLE EEWMGLLGYL SRISICKKKK DYCKMYRRHV RSRFYSIKDR NAR | |
Greater than 90% as determined by SDS-PAGE. |