missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nlrp3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Nlrp3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7856-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16001211

  • £2480.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_665826.1
Nlrp3 Protein
Research Use Only
Nlrp3
Recombinant Protein
E. coli
MTSVRCKLAQ YLEDLEDVDL KKFKMHLEDY PPEKGCIPVP RGQMEKADHL DLATLMIDFN GEEKAWAMAV WIFAAINRRD LWEKAKKDQP EWNDTCTSHS SMVCQEDSLE EEWMGLLGYL SRISICKKKK DYCKMYRRHV RSRFYSIKDR NAR
Greater than 90% as determined by SDS-PAGE.
216799
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers