missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nlrp3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nlrp3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7856-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_665826.1 | |
Nlrp3 Protein | |
Research Use Only | |
Nlrp3 | |
Recombinant Protein | |
E. coli | |
MTSVRCKLAQ YLEDLEDVDL KKFKMHLEDY PPEKGCIPVP RGQMEKADHL DLATLMIDFN GEEKAWAMAV WIFAAINRRD LWEKAKKDQP EWNDTCTSHS SMVCQEDSLE EEWMGLLGYL SRISICKKKK DYCKMYRRHV RSRFYSIKDR NAR | |
Greater than 90% as determined by SDS-PAGE. |
216799 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |