missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin reductase 2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin reductase 2, mitochondrial, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_006431.2 |
---|---|
Gene ID (Entrez) | 10587 |
Name | Thioredoxin reductase 2, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063231
|
enQuireBio™
QP8200-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083231
|
enQuireBio™
QP8200-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073231
|
enQuireBio™
QP8200-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093231
|
enQuireBio™
QP8200-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003241
|
enQuireBio™
QP8200-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013241
|
enQuireBio™
QP8200-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006431.2 | |
Thioredoxin reductase 2, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10587 | |
Research Use Only | |
TXNRD2 | |
Recombinant Protein | |
E. coli | |
QRDYDLLVVG GGSGGLACAK EAAQLGRKVA VVDYVEPSPQ GTRWGLGGTC VNVGCIPKKL MHQAALLGGL IQDAPNYGWE VAQPVPHDWR KMAEAVQNHV KSLNWGHRVQ LQDRKVKYFN IKASFVDEHT VCGVAKGGKE ILLSADHIII ATGGRPRYPT HIEGALEYGI TSDDIFWLKE SPGKTLVVGA SYVALECAGF LTGIGLDTTI MMRSIPLRGF DQQMSSMVIE HMASHGTRFL RGCAPSRVRR LPDGQLQVTW EDSTTGKEDT GTFDTVLWAI GRVPDTRSLN LEKAGVDTSP DTQKILVDSR EATSVPHIYA IGDVVEGRPE LTPIAIMAGR LLVQRLFGGS SDLMDYDNVP TTVFTPLEYG CVGLSEEEAV ARHGQEHVEV YHAHYKPLEF TVAGRDASQC YVKMVCLREP PQLVLGLHFL GPNAGEVTQG FALGIKCGAS YAQVMRTVGI HPTCSEEVVK LRISKRSGLD PTVTGCUG | |
Greater than 90% as determined by SDS-PAGE. |