missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thioredoxin reductase 2, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Thioredoxin reductase 2, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8200-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003241

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_006431.2
Thioredoxin reductase 2, mitochondrial Protein
Research Use Only
TXNRD2
Recombinant Protein
E. coli
QRDYDLLVVG GGSGGLACAK EAAQLGRKVA VVDYVEPSPQ GTRWGLGGTC VNVGCIPKKL MHQAALLGGL IQDAPNYGWE VAQPVPHDWR KMAEAVQNHV KSLNWGHRVQ LQDRKVKYFN IKASFVDEHT VCGVAKGGKE ILLSADHIII ATGGRPRYPT HIEGALEYGI TSDDIFWLKE SPGKTLVVGA SYVALECAGF LTGIGLDTTI MMRSIPLRGF DQQMSSMVIE HMASHGTRFL RGCAPSRVRR LPDGQLQVTW EDSTTGKEDT GTFDTVLWAI GRVPDTRSLN LEKAGVDTSP DTQKILVDSR EATSVPHIYA IGDVVEGRPE LTPIAIMAGR LLVQRLFGGS SDLMDYDNVP TTVFTPLEYG CVGLSEEEAV ARHGQEHVEV YHAHYKPLEF TVAGRDASQC YVKMVCLREP PQLVLGLHFL GPNAGEVTQG FALGIKCGAS YAQVMRTVGI HPTCSEEVVK LRISKRSGLD PTVTGCUG
Greater than 90% as determined by SDS-PAGE.
10587
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers