missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin reductase 2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin reductase 2, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8200-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006431.2 | |
Thioredoxin reductase 2, mitochondrial Protein | |
Research Use Only | |
TXNRD2 | |
Recombinant Protein | |
E. coli | |
QRDYDLLVVG GGSGGLACAK EAAQLGRKVA VVDYVEPSPQ GTRWGLGGTC VNVGCIPKKL MHQAALLGGL IQDAPNYGWE VAQPVPHDWR KMAEAVQNHV KSLNWGHRVQ LQDRKVKYFN IKASFVDEHT VCGVAKGGKE ILLSADHIII ATGGRPRYPT HIEGALEYGI TSDDIFWLKE SPGKTLVVGA SYVALECAGF LTGIGLDTTI MMRSIPLRGF DQQMSSMVIE HMASHGTRFL RGCAPSRVRR LPDGQLQVTW EDSTTGKEDT GTFDTVLWAI GRVPDTRSLN LEKAGVDTSP DTQKILVDSR EATSVPHIYA IGDVVEGRPE LTPIAIMAGR LLVQRLFGGS SDLMDYDNVP TTVFTPLEYG CVGLSEEEAV ARHGQEHVEV YHAHYKPLEF TVAGRDASQC YVKMVCLREP PQLVLGLHFL GPNAGEVTQG FALGIKCGAS YAQVMRTVGI HPTCSEEVVK LRISKRSGLD PTVTGCUG | |
Greater than 90% as determined by SDS-PAGE. |
10587 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |