missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNA binding protein fox-1 homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) RNA binding protein fox-1 homolog 2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001026865.1 |
---|---|
Gene ID (Entrez) | 23543 |
Name | RNA binding protein fox-1 homolog 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084350
|
enQuireBio™
QP6587-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004360
|
enQuireBio™
QP6587-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094350
|
enQuireBio™
QP6587-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014360
|
enQuireBio™
QP6587-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024360
|
enQuireBio™
QP6587-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034360
|
enQuireBio™
QP6587-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001026865.1 | |
RNA binding protein fox-1 homolog 2 Protein | |
Not determined. | |
Full Length of Isoform 10 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
23543 | |
Research Use Only | |
RBFOX2 | |
Recombinant Protein | |
E. coli | |
MEKKKMVTQG NQEPTTTPDA MVQPFTTIPF PPPPQNGIPT EYGVPHTQDY AGQTGEHNLT LYGSTQAHGE QSSNSPSTQN GSLTTEGGAQ TDGQQSQTQS SENSESKSTP KRLHVSNIPF RFRDPDLRQM FGQFGKILDV EIIFNERGSK GFGFVTFENS ADADRAREKL HGTVVEGRKI EVNNATARVM TNKKMVTPYA NGWKLSPVVG AVYGPELYAA SSFQADVSLG NDAAVPLSGR GGINTYIPLI SLPLVPGFPY PTAATTAAAF RGAHLRGRGR TVYGAVRAVP PTAIPAYPGV VYQDGFYGAD LYGGYAAYRY AQPATATAAT AAAAAAAAYS DGYGRVYTAD PYHALAPAAS YGVGAVASLY RGGYSRFAPY | |
Greater than 90% as determined by SDS-PAGE. |