missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RNA binding protein fox-1 homolog 2 Protein

A DNA sequence encoding the Homo sapiens (Human) RNA binding protein fox-1 homolog 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6587-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084350

  • £384.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001026865.1
RNA binding protein fox-1 homolog 2 Protein
Research Use Only
RBFOX2
Recombinant Protein
E. coli
MEKKKMVTQG NQEPTTTPDA MVQPFTTIPF PPPPQNGIPT EYGVPHTQDY AGQTGEHNLT LYGSTQAHGE QSSNSPSTQN GSLTTEGGAQ TDGQQSQTQS SENSESKSTP KRLHVSNIPF RFRDPDLRQM FGQFGKILDV EIIFNERGSK GFGFVTFENS ADADRAREKL HGTVVEGRKI EVNNATARVM TNKKMVTPYA NGWKLSPVVG AVYGPELYAA SSFQADVSLG NDAAVPLSGR GGINTYIPLI SLPLVPGFPY PTAATTAAAF RGAHLRGRGR TVYGAVRAVP PTAIPAYPGV VYQDGFYGAD LYGGYAAYRY AQPATATAAT AAAAAAAAYS DGYGRVYTAD PYHALAPAAS YGVGAVASLY RGGYSRFAPY
Greater than 90% as determined by SDS-PAGE.
23543
100 μg
Not determined.
Full Length of Isoform 10
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers