missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNA binding protein fox-1 homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) RNA binding protein fox-1 homolog 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6587-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001026865.1 | |
RNA binding protein fox-1 homolog 2 Protein | |
Research Use Only | |
RBFOX2 | |
Recombinant Protein | |
E. coli | |
MEKKKMVTQG NQEPTTTPDA MVQPFTTIPF PPPPQNGIPT EYGVPHTQDY AGQTGEHNLT LYGSTQAHGE QSSNSPSTQN GSLTTEGGAQ TDGQQSQTQS SENSESKSTP KRLHVSNIPF RFRDPDLRQM FGQFGKILDV EIIFNERGSK GFGFVTFENS ADADRAREKL HGTVVEGRKI EVNNATARVM TNKKMVTPYA NGWKLSPVVG AVYGPELYAA SSFQADVSLG NDAAVPLSGR GGINTYIPLI SLPLVPGFPY PTAATTAAAF RGAHLRGRGR TVYGAVRAVP PTAIPAYPGV VYQDGFYGAD LYGGYAAYRY AQPATATAAT AAAAAAAAYS DGYGRVYTAD PYHALAPAAS YGVGAVASLY RGGYSRFAPY | |
Greater than 90% as determined by SDS-PAGE. |
23543 | |
50 μg | |
Not determined. | |
Full Length of Isoform 10 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |