missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Major Vault Protein
A DNA sequence encoding the Homo sapiens (Human) Major Vault Protein, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001280133.1 |
---|---|
Gene ID (Entrez) | 9961 |
Name | Major Vault Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003030
|
enQuireBio™
QP6397-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023030
|
enQuireBio™
QP6397-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013030
|
enQuireBio™
QP6397-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033030
|
enQuireBio™
QP6397-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043030
|
enQuireBio™
QP6397-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053030
|
enQuireBio™
QP6397-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001280133.1 | |
Major Vault Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9961 | |
Research Use Only | |
MVP | |
Recombinant Protein | |
E. coli | |
ATEEFIIRIP PYHYIHVLDQ NSNVSRVEVG PKTYIRQDNE RVLFAPMRMV TVPPRHYCTV ANPVSRDAQG LVLFDVTGQV RLRHADLEIR LAQDPFPLYP GEVLEKDITP LQVVLPNTAL HLKALLDFED KDGDKVVAGD EWLFEGPGTY IPRKEVEVVE IIQATIIRQN QALRLRARKE CWDRDGKERV TGEEWLVTTV GAYLPAVFEE VLDLVDAVIL TEKTALHLRA RRNFRDFRGV SRRTGEEWLV TVQDTEAHVP DVHEEVLGVV PITTLGPHNY CVILDPVGPD GKNQLGQKRV VKGEKSFFLQ PGEQLEQGIQ DVYVLSEQQG LLLRALQPLE EGEDEEKVSH QAGDHWLIRG PLEYVPSAKV EVVEERQAIP LDENEGIYVQ DVKTGKVRAV IGSTYMLTQD EVLWEKELPP GVEELLNKGQ DPLADRGEKD TAKSLQPLAP RNKTRVVSYR VPHNAAVQVY DYREKRARVV FGPELVSLGP EEQFTVLSLS AGRPKRPHAR RALCLLLGPD FFTDVITIET ADHARLQLQL AYNWHFEVND RKDPQETAKL FSVPDFVGDA CKAIASRVRG AVASVTFDDF HKNSARIIRT AVFGFETSEA KGPDGMALPR PRDQAVFPQN GLVVSSVDVQ SVEPVDQRTR DALQRSVQLA IEITTNSQEA AAKHEAQRLE QEARGRLERQ KILDQSEAEK ARKELLELEA LSMAVESTGT AKAEAESRAE AARIEGEGSV LQAKLKAQAL AIETEAELQR VQKVRELELV YARAQLELEV SKAQQLAEVE VKKFKQMTEA IGPSTIRDLA VAGPEMQVKL LQSLGLKSTL ITDGSTPINL FNTAFGLLGM GPEGQPLGRR VASGPSPGEG ISPQSAQAPQ APGDNHVVPV LR | |
Greater than 90% as determined by SDS-PAGE. |