missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Major Vault Protein

A DNA sequence encoding the Homo sapiens (Human) Major Vault Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6397-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001280133.1
Major Vault Protein
Research Use Only
MVP
Recombinant Protein
E. coli
ATEEFIIRIP PYHYIHVLDQ NSNVSRVEVG PKTYIRQDNE RVLFAPMRMV TVPPRHYCTV ANPVSRDAQG LVLFDVTGQV RLRHADLEIR LAQDPFPLYP GEVLEKDITP LQVVLPNTAL HLKALLDFED KDGDKVVAGD EWLFEGPGTY IPRKEVEVVE IIQATIIRQN QALRLRARKE CWDRDGKERV TGEEWLVTTV GAYLPAVFEE VLDLVDAVIL TEKTALHLRA RRNFRDFRGV SRRTGEEWLV TVQDTEAHVP DVHEEVLGVV PITTLGPHNY CVILDPVGPD GKNQLGQKRV VKGEKSFFLQ PGEQLEQGIQ DVYVLSEQQG LLLRALQPLE EGEDEEKVSH QAGDHWLIRG PLEYVPSAKV EVVEERQAIP LDENEGIYVQ DVKTGKVRAV IGSTYMLTQD EVLWEKELPP GVEELLNKGQ DPLADRGEKD TAKSLQPLAP RNKTRVVSYR VPHNAAVQVY DYREKRARVV FGPELVSLGP EEQFTVLSLS AGRPKRPHAR RALCLLLGPD FFTDVITIET ADHARLQLQL AYNWHFEVND RKDPQETAKL FSVPDFVGDA CKAIASRVRG AVASVTFDDF HKNSARIIRT AVFGFETSEA KGPDGMALPR PRDQAVFPQN GLVVSSVDVQ SVEPVDQRTR DALQRSVQLA IEITTNSQEA AAKHEAQRLE QEARGRLERQ KILDQSEAEK ARKELLELEA LSMAVESTGT AKAEAESRAE AARIEGEGSV LQAKLKAQAL AIETEAELQR VQKVRELELV YARAQLELEV SKAQQLAEVE VKKFKQMTEA IGPSTIRDLA VAGPEMQVKL LQSLGLKSTL ITDGSTPINL FNTAFGLLGM GPEGQPLGRR VASGPSPGEG ISPQSAQAPQ APGDNHVVPV LR
Greater than 90% as determined by SDS-PAGE.
9961
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers