missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Major Vault Protein
A DNA sequence encoding the Homo sapiens (Human) Major Vault Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6397-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001280133.1 | |
Major Vault Protein | |
Research Use Only | |
MVP | |
Recombinant Protein | |
E. coli | |
ATEEFIIRIP PYHYIHVLDQ NSNVSRVEVG PKTYIRQDNE RVLFAPMRMV TVPPRHYCTV ANPVSRDAQG LVLFDVTGQV RLRHADLEIR LAQDPFPLYP GEVLEKDITP LQVVLPNTAL HLKALLDFED KDGDKVVAGD EWLFEGPGTY IPRKEVEVVE IIQATIIRQN QALRLRARKE CWDRDGKERV TGEEWLVTTV GAYLPAVFEE VLDLVDAVIL TEKTALHLRA RRNFRDFRGV SRRTGEEWLV TVQDTEAHVP DVHEEVLGVV PITTLGPHNY CVILDPVGPD GKNQLGQKRV VKGEKSFFLQ PGEQLEQGIQ DVYVLSEQQG LLLRALQPLE EGEDEEKVSH QAGDHWLIRG PLEYVPSAKV EVVEERQAIP LDENEGIYVQ DVKTGKVRAV IGSTYMLTQD EVLWEKELPP GVEELLNKGQ DPLADRGEKD TAKSLQPLAP RNKTRVVSYR VPHNAAVQVY DYREKRARVV FGPELVSLGP EEQFTVLSLS AGRPKRPHAR RALCLLLGPD FFTDVITIET ADHARLQLQL AYNWHFEVND RKDPQETAKL FSVPDFVGDA CKAIASRVRG AVASVTFDDF HKNSARIIRT AVFGFETSEA KGPDGMALPR PRDQAVFPQN GLVVSSVDVQ SVEPVDQRTR DALQRSVQLA IEITTNSQEA AAKHEAQRLE QEARGRLERQ KILDQSEAEK ARKELLELEA LSMAVESTGT AKAEAESRAE AARIEGEGSV LQAKLKAQAL AIETEAELQR VQKVRELELV YARAQLELEV SKAQQLAEVE VKKFKQMTEA IGPSTIRDLA VAGPEMQVKL LQSLGLKSTL ITDGSTPINL FNTAFGLLGM GPEGQPLGRR VASGPSPGEG ISPQSAQAPQ APGDNHVVPV LR | |
Greater than 90% as determined by SDS-PAGE. |
9961 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |