missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LGR5 Protein
A DNA sequence encoding the Homo sapiens (Human) LGR5, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_001264155.1 |
---|---|
Gene ID (Entrez) | 8549 |
Name | LGR5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047911
|
enQuireBio™
QP9247-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087911
|
enQuireBio™
QP9247-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057911
|
enQuireBio™
QP9247-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077911
|
enQuireBio™
QP9247-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097911
|
enQuireBio™
QP9247-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067911
|
enQuireBio™
QP9247-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001264155.1 | |
LGR5 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
8549 | |
Research Use Only | |
LGR5 | |
Recombinant Protein | |
Yeast | |
GSSPRSGVLL RGCPTHCHCE PDGRMLLRVD CSDLGLSELP SNLSVFTSYL DLSMNNISQL LPNPLPSLRF LEELRLAGNA LTYIPKGAFT GLYSLKVLML QNNQLRHVPT EALQNLRSLQ SLRLDANHIS YVPPSCFSGL HSLRHLWLDD NALTEIPVQA FRSLSALQAM TLALNKIHHI PDYAFGNLSS LVVLHLHNNR IHSLGKKCFD GLHSLETLDL NYNNLDEFPT AIRTLSNLKE LGFHSNNIRS IPEKAFVGNP SLITIHFYDN PIQFVGRSAF QHLPELRTLT LNGASQITEF PDLTGTANLE SLTLTGAQIS SLPQTVCNQL PNLQVLDLSY NLLEDLPSFS VCQKLQKIDL RHNEIYEIKV DTFQQLLSLR SLNLAWNKIA IIHPNAFSTL PSLIKLDLSS NLLSSFPITG LHGLTHLKLT GNHALQSLIS SENFPELKVI EMPYAYQCCA FGVCENAYKI SNQWNKGDNS SMDDLHKKDA GMFQAQDERD LEDFLLDFEE DLKALHSVQC SPSPGPFKPC EHLLDGWLIR | |
Greater than 80% as determined by SDS-PAGE. |