missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LGR5 Protein

A DNA sequence encoding the Homo sapiens (Human) LGR5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9247-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087911

  • £1265.00 / 500µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001264155.1
LGR5 Protein
Research Use Only
LGR5
Recombinant Protein
Yeast
GSSPRSGVLL RGCPTHCHCE PDGRMLLRVD CSDLGLSELP SNLSVFTSYL DLSMNNISQL LPNPLPSLRF LEELRLAGNA LTYIPKGAFT GLYSLKVLML QNNQLRHVPT EALQNLRSLQ SLRLDANHIS YVPPSCFSGL HSLRHLWLDD NALTEIPVQA FRSLSALQAM TLALNKIHHI PDYAFGNLSS LVVLHLHNNR IHSLGKKCFD GLHSLETLDL NYNNLDEFPT AIRTLSNLKE LGFHSNNIRS IPEKAFVGNP SLITIHFYDN PIQFVGRSAF QHLPELRTLT LNGASQITEF PDLTGTANLE SLTLTGAQIS SLPQTVCNQL PNLQVLDLSY NLLEDLPSFS VCQKLQKIDL RHNEIYEIKV DTFQQLLSLR SLNLAWNKIA IIHPNAFSTL PSLIKLDLSS NLLSSFPITG LHGLTHLKLT GNHALQSLIS SENFPELKVI EMPYAYQCCA FGVCENAYKI SNQWNKGDNS SMDDLHKKDA GMFQAQDERD LEDFLLDFEE DLKALHSVQC SPSPGPFKPC EHLLDGWLIR
Greater than 80% as determined by SDS-PAGE.
8549
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers