missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LGR5 Protein
A DNA sequence encoding the Homo sapiens (Human) LGR5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9247-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001264155.1 | |
LGR5 Protein | |
Research Use Only | |
LGR5 | |
Recombinant Protein | |
Yeast | |
GSSPRSGVLL RGCPTHCHCE PDGRMLLRVD CSDLGLSELP SNLSVFTSYL DLSMNNISQL LPNPLPSLRF LEELRLAGNA LTYIPKGAFT GLYSLKVLML QNNQLRHVPT EALQNLRSLQ SLRLDANHIS YVPPSCFSGL HSLRHLWLDD NALTEIPVQA FRSLSALQAM TLALNKIHHI PDYAFGNLSS LVVLHLHNNR IHSLGKKCFD GLHSLETLDL NYNNLDEFPT AIRTLSNLKE LGFHSNNIRS IPEKAFVGNP SLITIHFYDN PIQFVGRSAF QHLPELRTLT LNGASQITEF PDLTGTANLE SLTLTGAQIS SLPQTVCNQL PNLQVLDLSY NLLEDLPSFS VCQKLQKIDL RHNEIYEIKV DTFQQLLSLR SLNLAWNKIA IIHPNAFSTL PSLIKLDLSS NLLSSFPITG LHGLTHLKLT GNHALQSLIS SENFPELKVI EMPYAYQCCA FGVCENAYKI SNQWNKGDNS SMDDLHKKDA GMFQAQDERD LEDFLLDFEE DLKALHSVQC SPSPGPFKPC EHLLDGWLIR | |
Greater than 80% as determined by SDS-PAGE. |
8549 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |