missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNG / Interferon Gamma Protein
A DNA sequence encoding the Homo sapiens (Human) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
£261.83 - £2965.00
Specifications
Accession Number | NP_000610.2 |
---|---|
Gene ID (Entrez) | 3458 |
Name | IFNG / Interferon Gamma Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16095211
|
enQuireBio™
QP8548-YE-100UG |
100 μg |
£670.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16015221
|
enQuireBio™
QP8548-YE-1MG |
1 mg |
£2965.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005221
|
enQuireBio™
QP8548-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025221
|
enQuireBio™
QP8548-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035221
|
enQuireBio™
QP8548-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045221
|
enQuireBio™
QP8548-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000610.2 | |
IFNG / Interferon Gamma Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3458 | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
Yeast | |
QDPYVKEAEN LKKYFNAGHS DVADNGTLFL GILKNWKEES DRKIMQSQIV SFYFKLFKNF KDDQSIQKSV ETIKEDMNVK FFNSNKKKRD DFEKLTNYSV TDLNVQRKAI HELIQVMAEL SPAAKTGKRK RSQMLFRG | |
Greater than 90% as determined by SDS-PAGE. |