missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNG / Interferon Gamma Protein
A DNA sequence encoding the Homo sapiens (Human) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8548-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000610.2 | |
IFNG / Interferon Gamma Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
Yeast | |
QDPYVKEAEN LKKYFNAGHS DVADNGTLFL GILKNWKEES DRKIMQSQIV SFYFKLFKNF KDDQSIQKSV ETIKEDMNVK FFNSNKKKRD DFEKLTNYSV TDLNVQRKAI HELIQVMAEL SPAAKTGKRK RSQMLFRG | |
Greater than 90% as determined by SDS-PAGE. |
3458 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |