missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFNG / Interferon Gamma Protein

A DNA sequence encoding the Homo sapiens (Human) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8548-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095211

  • £670.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000610.2
IFNG / Interferon Gamma Protein
Research Use Only
IFNG
Recombinant Protein
Yeast
QDPYVKEAEN LKKYFNAGHS DVADNGTLFL GILKNWKEES DRKIMQSQIV SFYFKLFKNF KDDQSIQKSV ETIKEDMNVK FFNSNKKKRD DFEKLTNYSV TDLNVQRKAI HELIQVMAEL SPAAKTGKRK RSQMLFRG
Greater than 90% as determined by SDS-PAGE.
3458
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers