missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF11 Protein
Recombinant Protein
£80.80 - £3965.00
Specifications
Accession Number | NP_005802.1 |
---|---|
Gene ID (Entrez) | 10220 |
Name | GDF11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975999
|
enQuireBio™
QP5369-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955999
|
enQuireBio™
QP5369-20UG |
20 μg |
£191.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935999
|
enQuireBio™
QP5369-100UG |
100 μg |
£525.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965999
|
enQuireBio™
QP5369-500UG |
500 μg |
£2425.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945999
|
enQuireBio™
QP5369-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_005802.1 | |
GDF11 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Alkaline phosphatase activity was demonstrated in ATDC5 cells significantly at concentrations less than 100 ng/ml. Typical ED50 is in the range of 10 ng/ml. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
10220 | |
Research Use Only | |
GDF11 | |
Recombinant Protein | |
E. coli | |
NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS | |
Greater than 95% as determined by SDS-PAGE. |