missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF11 Protein
Recombinant Protein
Brand: enQuireBio™ QP5369-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005802.1 | |
GDF11 Protein | |
Research Use Only | |
GDF11 | |
Recombinant Protein | |
E. coli | |
NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS | |
Greater than 95% as determined by SDS-PAGE. |
10220 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Alkaline phosphatase activity was demonstrated in ATDC5 cells significantly at concentrations less than 100 ng/ml. Typical ED50 is in the range of 10 ng/ml. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |