missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF11 Protein
Specifications
Specifications
| Accession Number | NP_005802.1 |
| Gene ID (Entrez) | 10220 |
| Name | GDF11 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | GDF11 |
| Biological Activity | Alkaline phosphatase activity was demonstrated in ATDC5 cells significantly at concentrations less than 100 ng/ml. Typical ED50 is in the range of 10 ng/ml. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?