missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDF11 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5369-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965999

  • £2425.00 / 500µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005802.1
GDF11 Protein
Research Use Only
GDF11
Recombinant Protein
E. coli
NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
Greater than 95% as determined by SDS-PAGE.
10220
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Alkaline phosphatase activity was demonstrated in ATDC5 cells significantly at concentrations less than 100 ng/ml. Typical ED50 is in the range of 10 ng/ml.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers