missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynactin subunit 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 3, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001268354.1 |
---|---|
Gene ID (Entrez) | 11258 |
Name | Dynactin subunit 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958669
|
enQuireBio™
QP5918-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978669
|
enQuireBio™
QP5918-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968669
|
enQuireBio™
QP5918-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988669
|
enQuireBio™
QP5918-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998669
|
enQuireBio™
QP5918-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908679
|
enQuireBio™
QP5918-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001268354.1 | |
Dynactin subunit 3 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11258 | |
Research Use Only | |
DCTN3 | |
Recombinant Protein | |
E. coli | |
AGLTDLQRLQ ARVEELERWV YGPGGARGSR KVADGLVKVQ VALGNISSKR ERVKILYKKI EDLIKYLDPE YIDRIAIPDA SKLQFILAEE QFILSQVALL EQVNALVPML DSAHIKAVPE HAARLQRLAQ IHIQQQAPWG VGVRDEAGSL VEDVGFAQFL SVLHFGPTGP VCGNH | |
Greater than 90% as determined by SDS-PAGE. |