missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynactin subunit 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5918-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001268354.1 | |
Dynactin subunit 3 Protein | |
Research Use Only | |
DCTN3 | |
Recombinant Protein | |
E. coli | |
AGLTDLQRLQ ARVEELERWV YGPGGARGSR KVADGLVKVQ VALGNISSKR ERVKILYKKI EDLIKYLDPE YIDRIAIPDA SKLQFILAEE QFILSQVALL EQVNALVPML DSAHIKAVPE HAARLQRLAQ IHIQQQAPWG VGVRDEAGSL VEDVGFAQFL SVLHFGPTGP VCGNH | |
Greater than 90% as determined by SDS-PAGE. |
11258 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |