missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dynactin subunit 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Dynactin subunit 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5918-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978669

  • £1735.00 / 1mg
Estimated Shipment: 03-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001268354.1
Dynactin subunit 3 Protein
Research Use Only
DCTN3
Recombinant Protein
E. coli
AGLTDLQRLQ ARVEELERWV YGPGGARGSR KVADGLVKVQ VALGNISSKR ERVKILYKKI EDLIKYLDPE YIDRIAIPDA SKLQFILAEE QFILSQVALL EQVNALVPML DSAHIKAVPE HAARLQRLAQ IHIQQQAPWG VGVRDEAGSL VEDVGFAQFL SVLHFGPTGP VCGNH
Greater than 90% as determined by SDS-PAGE.
11258
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers