missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA dC->dU-editing enzyme APOBEC-3C Protein
A DNA sequence encoding the Homo sapiens (Human) DNA dC->dU-editing enzyme APOBEC-3C, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_055323.2 |
---|---|
Gene ID (Entrez) | 27350 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | APOBEC3C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003381
|
enQuireBio™
QP8223-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023381
|
enQuireBio™
QP8223-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013381
|
enQuireBio™
QP8223-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033381
|
enQuireBio™
QP8223-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043381
|
enQuireBio™
QP8223-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053381
|
enQuireBio™
QP8223-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055323.2 | |
Research Use Only | |
APOBEC3C | |
Recombinant Protein | |
E. coli | |
MNPQIRNPMK AMYPGTFYFQ FKNLWEANDR DETWLCFTVE GIKRRSVVSW KTGVFRNQVD SETHCHAERC FLSWFCDDIL SPNTKYQVTW YTSWSPCPDC AGEVAEFLAR HSNVNLTIFT ARLYYFQYPC YQEGLRSLSQ EGVAVEIMDY EDFKYCWENF VYNDNEPFKP WKGLKTNFRL LKRRLRESLQ | |
Greater than 80% as determined by SDS-PAGE. |
27350 | |
Not determined. | |
Full Length of BC011739 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |