missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA dC->dU-editing enzyme APOBEC-3C Protein

A DNA sequence encoding the Homo sapiens (Human) DNA dC->dU-editing enzyme APOBEC-3C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8223-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023381

  • £2480.00 / 1mg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_055323.2
DNA dC->dU-editing enzyme APOBEC-3C Protein
Research Use Only
APOBEC3C
Recombinant Protein
E. coli
MNPQIRNPMK AMYPGTFYFQ FKNLWEANDR DETWLCFTVE GIKRRSVVSW KTGVFRNQVD SETHCHAERC FLSWFCDDIL SPNTKYQVTW YTSWSPCPDC AGEVAEFLAR HSNVNLTIFT ARLYYFQYPC YQEGLRSLSQ EGVAVEIMDY EDFKYCWENF VYNDNEPFKP WKGLKTNFRL LKRRLRESLQ
Greater than 80% as determined by SDS-PAGE.
27350
1 mg
Not determined.
Full Length of BC011739
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers