missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA dC->dU-editing enzyme APOBEC-3C Protein
A DNA sequence encoding the Homo sapiens (Human) DNA dC->dU-editing enzyme APOBEC-3C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8223-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055323.2 | |
DNA dC->dU-editing enzyme APOBEC-3C Protein | |
Research Use Only | |
APOBEC3C | |
Recombinant Protein | |
E. coli | |
MNPQIRNPMK AMYPGTFYFQ FKNLWEANDR DETWLCFTVE GIKRRSVVSW KTGVFRNQVD SETHCHAERC FLSWFCDDIL SPNTKYQVTW YTSWSPCPDC AGEVAEFLAR HSNVNLTIFT ARLYYFQYPC YQEGLRSLSQ EGVAVEIMDY EDFKYCWENF VYNDNEPFKP WKGLKTNFRL LKRRLRESLQ | |
Greater than 80% as determined by SDS-PAGE. |
27350 | |
50 μg | |
Not determined. | |
Full Length of BC011739 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |