missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cathepsin K Protein
A DNA sequence encoding the Homo sapiens (Human) Cathepsin K, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_000387.1 |
---|---|
Gene ID (Entrez) | 1513 |
Name | Cathepsin K Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15998479
|
enQuireBio™
QP5889-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938489
|
enQuireBio™
QP5889-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15908489
|
enQuireBio™
QP5889-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15928489
|
enQuireBio™
QP5889-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15948489
|
enQuireBio™
QP5889-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15918489
|
enQuireBio™
QP5889-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_000387.1 | |
Cathepsin K Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1513 | |
Research Use Only | |
CTSK | |
Recombinant Protein | |
Yeast | |
APDSVDYRKK GYVTPVKNQG QCGSCWAFSS VGALEGQLKK KTGKLLNLSP QNLVDCVSEN DGCGGGYMTN AFQYVQKNRG IDSEDAYPYV GQEESCMYNP TGKAAKCRGY REIPEGNEKA LKRAVARVGP VSVAIDASLT SFQFYSKGVY YDESCNSDNL NHAVLAVGYG IQKGNKHWII KNSWGENWGN KGYILMARNK NNACGIANLA SFPKM | |
Greater than 90% as determined by SDS-PAGE. |