missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cathepsin K Protein
A DNA sequence encoding the Homo sapiens (Human) Cathepsin K, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5889-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000387.1 | |
Cathepsin K Protein | |
Research Use Only | |
CTSK | |
Recombinant Protein | |
Yeast | |
APDSVDYRKK GYVTPVKNQG QCGSCWAFSS VGALEGQLKK KTGKLLNLSP QNLVDCVSEN DGCGGGYMTN AFQYVQKNRG IDSEDAYPYV GQEESCMYNP TGKAAKCRGY REIPEGNEKA LKRAVARVGP VSVAIDASLT SFQFYSKGVY YDESCNSDNL NHAVLAVGYG IQKGNKHWII KNSWGENWGN KGYILMARNK NNACGIANLA SFPKM | |
Greater than 90% as determined by SDS-PAGE. |
1513 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |